missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) isopropoxide, 97+%

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids

Manufacturer:  Alfa Aesar™ 7711522

 View more versions of this product

Catalog No. AA7711522



Specifications

Specifications

Titanium (IV) isopropoxide
C12H28O4Ti
MFCD00008871
3679474
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
VXUYXOFXAQZZMF-UHFFFAOYSA-N
propan-2-olate;titanium(4+)
11026
Liquid
0.955g/mL
Moisture Sensitive
Colorless to Yellow
100g
546-68-9
Ti[OCH(CH3)2]4
UN2413
14,9480
Soluble in anhydrous ethanol, ether, benzene and chloroform; Decomposes in water
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]
284.219
284.23
≥97%
1.4640
232°C
16°C to 20°C
46°C (115°F)

Chemical Identifiers

546-68-9
284.219
VXUYXOFXAQZZMF-UHFFFAOYSA-N
11026
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]
C12H28O4Ti
MFCD00008871
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
propan-2-olate;titanium(4+)
Safety and Handling

Safety and Handling

GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAPROPYLORTHOTITANATE

RTECSNumber : NT8060000

TSCA : TSCA

SDS Documents