missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) isobutoxide

Manufacturer:  Alfa Aesar™ 8945530

 View more versions of this product

Catalog No. AA8945530



Specifications

Specifications

Titanium (IV) isobutoxide
C16H36O4Si
MFCD00015045
QKCMIPQMYBKEPP-UHFFFAOYSA-N
tetrakis(2-methylpropyl) silicate
69626
Melt
1.4950
296°C
250g
681-98-1
Ti[(CH3)2CHCH2O]4
tetrakis 2-methylpropyl orthosilicate, titanium iv isobutoxide, tetrakis 2-methylpropyl silicate, tetraisobutyl orthosilicate, silane, tetraisobutoxy, silicic acid tetraisobutyl ester, orthosilicic acid tetrakis 2-methylpropyl ester, silicic acid h4sio4 ,tetrakis 2-methylpropyl ester
CC(C)CO[Si](OCC(C)C)(OCC(C)C)OCC(C)C
320.545
340.34
1.02g/mL
Moisture Sensitive
Yellow
77°C (171°F)

Chemical Identifiers

681-98-1
320.545
QKCMIPQMYBKEPP-UHFFFAOYSA-N
69626
CC(C)CO[Si](OCC(C)C)(OCC(C)C)OCC(C)C
C16H36O4Si
MFCD00015045
tetrakis 2-methylpropyl orthosilicate, titanium iv isobutoxide, tetrakis 2-methylpropyl silicate, tetraisobutyl orthosilicate, silane, tetraisobutoxy, silicic acid tetraisobutyl ester, orthosilicic acid tetrakis 2-methylpropyl ester, silicic acid h4sio4 ,tetrakis 2-methylpropyl ester
tetrakis(2-methylpropyl) silicate
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents