missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) iodide 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC391880250

 View more versions of this product

Catalog No. AC391880250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94% (ex Ti)
Fused Solid
Purple to black
Titanium tetraiodide
Titanium(IV) iodide
MFCD00043077
7720-83-4
555.48
25g
Glass bottle
95%
I4Ti
TiI4

Chemical Identifiers

95%
TiI4
MFCD00043077
Titanium(IV) iodide
I4Ti
555.48
Titanium tetraiodide
≥94% (ex Ti)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger