Please login to your online account to display your discounted pricing

Titanium(IV) iodide 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Titanium(IV) iodide
Assay ≥94% (ex Ti)
Assay Percent Range 95%
Molecular Formula I4Ti
Linear Formula TiI4
Formula Weight 555.48
MDL Number MFCD00043077
Synonym Titanium tetraiodide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391880250

  SDS
thermo fisher scientific
AC391880250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94% (ex Ti)
Physical Form Fused Solid
Formula Weight 555.48
Quantity 25g
Color Purple to black
Chemical Name or Material Titanium(IV) iodide
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula I4Ti
MDL Number MFCD00043077
Synonym Titanium tetraiodide
Linear Formula TiI4
CAS 7720-83-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Lewis acid

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.