Please login to your online account to display your discounted pricing

Titanium(IV) fluoride, 99%, ACROS Organics™


Quantity 10g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  308430100

 View more versions of this product

Catalog No. AC308430100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (ex F)
Physical Form Powder and/or Chunks
Formula Weight 123.89
Quantity 10g
Color White
Merck Index 15, 9635
Chemical Name or Material Titanium(IV) fluoride
Packaging Plastic bottle
Assay Percent Range 99%
Molecular Formula TiF4
Linear Formula Ti F4
MDL Number MFCD00042545
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 7783-63-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

Specifications

Chemical Name or Material Titanium(IV) fluoride
Assay ≥98.5% (ex F)
Assay Percent Range 99%
Molecular Formula TiF4
Linear Formula Ti F4
Formula Weight 123.89
MDL Number MFCD00042545
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents