missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) ethoxide, 85%, Tech., contains 5-15% isopropanol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 312540500

 View more versions of this product

Catalog No. AC312540500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Titanium(IV) ethoxide
3087-36-3,67-63-0
1.0800g/mL
29°C
85% min
C8H20O4Ti
Ti(OC2H5)4
50 g
1.08
Solubility in water: hydrolysis.
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
228.15
228.15
Technical
Contains 5-15% Isopropanol
Colorless to Yellow
150.0°C to 152.0°C (10.0 mmHg)
Authentic
Glass bottle
1.5033 to 1.5053
MFCD00009071
01,III,1338
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
JMXKSZRRTHPKDL-UHFFFAOYSA-N
ethanolate;titanium(4+)
76524
85%
Liquid

Chemical Identifiers

3087-36-3
228.15
JMXKSZRRTHPKDL-UHFFFAOYSA-N
76524
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
C8H20O4Ti
MFCD00009071
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
ethanolate;titanium(4+)
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapour.
Causes serious eye irritation.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-410-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only