Please login to your online account to display your discounted pricing

Titanium(IV) ethoxide, 33-35% TiO2, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  223762500

 View more versions of this product

Catalog No. AC223762500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 228.15
Physical Form Liquid
Quantity 250g
Color Clear colorless to yellow
Assay 33 to 35% (TiO2)
Density 1.0900g/mL
Chemical Name or Material Titanium(IV) ethoxide
Linear Formula Ti(OC2H5)4
CAS 3087-36-3
Infrared Spectrum Authentic
Refractive Index 1.5050 to 1.5070 (20°C, 589nm)
Packaging Glass bottle
Molecular Formula C8H20O4Ti

Specifications

Chemical Name or Material Titanium(IV) ethoxide
Assay 33 to 35% (TiO2)
Molecular Formula C8H20O4Ti
Linear Formula Ti(OC2H5)4
Formula Weight 228.15