Learn More
Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 223760010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Lewis acidSpecifications
Titanium(IV) ethoxide | |
1.0900g/mL | |
33°C | |
Glass bottle | |
1.5050 to 1.5070 | |
1kg | |
1.09 | |
Solubility in water: hydrolysis. | |
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4] | |
228.15 | |
60-80 mPa.s (20°C) | |
33-35% TiO2 |
3087-36-3 | |
150.0°C to 152.0°C (10.0mmHg) | |
Authentic | |
C8H20O4Ti | |
Ti(OC2H5)4 | |
01,III,1338 | |
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate | |
JMXKSZRRTHPKDL-UHFFFAOYSA-N | |
ethanolate;titanium(4+) | |
76524 | |
228.15 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
Wash f
GHS Signal Word: Warning
EINECSNumber : 221-410-8
RUO â Research Use Only