missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) chloride, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 2297914

 View more versions of this product

Catalog No. AA2297914



Specifications

Specifications

Titanium(IV) chloride
99.99% (metals basis)
MFCD00011267
XJDNKRIXUMDJCW-UHFFFAOYSA-J
titanium(4+);tetrachloride
160960
Liquid
7550-45-0
Cl4Ti
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
189.667
189.69
25g

Chemical Identifiers

7550-45-0
189.667
XJDNKRIXUMDJCW-UHFFFAOYSA-J
160960
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Cl4Ti
MFCD00011267
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
titanium(4+);tetrachloride
Safety and Handling

Safety and Handling

EINECSNumber : 231-441-9

SDS Documents