missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Titanium(IV) chloride, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 377748000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Titanium(IV) chloride | |
7550-45-0,75-09-2 | |
Solution | |
Cl4Ti | |
15, 9634 | |
Solubility in water: reacts | |
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4] | |
189.71 | |
189.71 | |
AcroSeal™ Glass Bottle | |
1.3650g/mL |
1M Solution in Dichloromethane | |
Brown to Yellow | |
800 mL | |
MFCD00011267 | |
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4 | |
XJDNKRIXUMDJCW-UHFFFAOYSA-J | |
titanium(4+);tetrachloride | |
160960 | |
0.95 to 1.10M | |
1.365 |
RUO – Research Use Only