missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) 2-ethylhexanoate, 97%

Manufacturer:  Alfa Aesar™ 4460222

 View more versions of this product

Catalog No. AA4460222



Specifications

Specifications

Titanium (IV) 2-ethylhexanoate
100g
C32H60O8Ti
2-ethylhexanoate; titanium 4+
GDKAXSGMPFSRJY-UHFFFAOYSA-J
2-ethylhexanoate;titanium(4+)
6451578
Liquid
Odorless
3645-34-9
MFCD05663781
Immiscible with water. Miscible with ethyl acetate.
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ti+4]
620.691
623.75
97%

Chemical Identifiers

3645-34-9
620.691
GDKAXSGMPFSRJY-UHFFFAOYSA-J
6451578
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ti+4]
C32H60O8Ti
MFCD05663781
2-ethylhexanoate; titanium 4+
2-ethylhexanoate;titanium(4+)
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H361d

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents