Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) 2-ethylhexanoate, 97%

$189.34 - $717.31

Chemical Identifiers

Chemical Name or Material Titanium (IV) 2-ethylhexanoate
CAS 3645-34-9
Molecular Formula C32H60O8Ti
Formula Weight 623.75
MDL Number MFCD05663781
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4460222

  SDS
alfa aesar™
4460222
100g Each for $189.34

AA4460236

  SDS
alfa aesar™
4460236
500g Each for $717.31
Description & Specifications

Specifications

Formula Weight 623.75
Chemical Name or Material Titanium (IV) 2-ethylhexanoate
TSCA TSCA
Physical Form Liquid
Assay 97%
Molecular Formula C32H60O8Ti
MDL Number MFCD05663781
CAS 3645-34-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Health Hazard 3 GHS P Statement
P280h
Wear protective gloves/clothing.
Quantity 100g
Odor Odorless
Safety and Handling

Warning

GHS P Statement
P280h
Wear protective gloves/clothing.

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.