missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(III) oxide, 99.8% (metals basis)

$258.76 - $1,155.58

Chemical Identifiers

CAS 1344-54-3
Molecular Formula O3Ti2
Molecular Weight (g/mol) 143.731
MDL Number MFCD00036278
InChI Key GQUJEMVIKWQAEH-UHFFFAOYSA-N
Synonym oxo oxotitaniooxy titanium, oxo-oxotitaniooxy titanium, ti2o3, ti2o3 nanoparticle, titanium iii oxide, titanium iii oxide nanoparticle, titanium iii oxide trace metals basis 50g, titanium oxide ti2o3, titanium sesquioxide, titatnum iii oxide
PubChem CID 123111
IUPAC Name oxo(oxotitaniooxy)titanium
SMILES O=[Ti]O[Ti]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA7712718
View Documents
Alfa Aesar™
7712718
50g Each for $258.76
 
AA7712730
View Documents
Alfa Aesar™
7712730
250g Each for $1,155.58
 
Specifications

Chemical Identifiers

1344-54-3
143.731
GQUJEMVIKWQAEH-UHFFFAOYSA-N
123111
O=[Ti]O[Ti]=O
O3Ti2
MFCD00036278
oxo oxotitaniooxy titanium, oxo-oxotitaniooxy titanium, ti2o3, ti2o3 nanoparticle, titanium iii oxide, titanium iii oxide nanoparticle, titanium iii oxide trace metals basis 50g, titanium oxide ti2o3, titanium sesquioxide, titatnum iii oxide
oxo(oxotitaniooxy)titanium

Specifications

1344-54-3
MFCD00036278
Insoluble in water.
O=[Ti]O[Ti]=O
143.731
143.8
99.8%
−325 Mesh
2130°C (decomposition)
Titanium (III) oxide
O3Ti2
oxo oxotitaniooxy titanium, oxo-oxotitaniooxy titanium, ti2o3, ti2o3 nanoparticle, titanium iii oxide, titanium iii oxide nanoparticle, titanium iii oxide trace metals basis 50g, titanium oxide ti2o3, titanium sesquioxide, titatnum iii oxide
GQUJEMVIKWQAEH-UHFFFAOYSA-N
oxo(oxotitaniooxy)titanium
123111
-325 Mesh Powder
4.6 g/mL
Odorless
50g
(metals basis)
Safety and Handling

Safety and Handling

EINECSNumber : 215-697-9

TSCA : Yes

Recommended Storage : Ambient temperatures