missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(III) oxide, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 7712718

 View more versions of this product

Catalog No. AA7712718



Specifications

Specifications

Titanium (III) oxide
(metals basis)
MFCD00036278
Insoluble in water.
O=[Ti]O[Ti]=O
143.731
143.8
99.8%
−325 Mesh
2130°C (decomposition)
1344-54-3
O3Ti2
oxo oxotitaniooxy titanium, oxo-oxotitaniooxy titanium, ti2o3, ti2o3 nanoparticle, titanium iii oxide, titanium iii oxide nanoparticle, titanium iii oxide trace metals basis 50g, titanium oxide ti2o3, titanium sesquioxide, titatnum iii oxide
GQUJEMVIKWQAEH-UHFFFAOYSA-N
oxo(oxotitaniooxy)titanium
123111
-325 Mesh Powder
4.6 g/mL
Odorless
50g

Chemical Identifiers

1344-54-3
143.731
GQUJEMVIKWQAEH-UHFFFAOYSA-N
123111
O=[Ti]O[Ti]=O
O3Ti2
MFCD00036278
oxo oxotitaniooxy titanium, oxo-oxotitaniooxy titanium, ti2o3, ti2o3 nanoparticle, titanium iii oxide, titanium iii oxide nanoparticle, titanium iii oxide trace metals basis 50g, titanium oxide ti2o3, titanium sesquioxide, titatnum iii oxide
oxo(oxotitaniooxy)titanium
Safety and Handling

Safety and Handling

EINECSNumber : 215-697-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents