Please login to your online account to display your discounted pricing

Titanium(III) fluoride, 98%, ACROS Organics™


Quantity 1g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC384520010

 View more versions of this product

Catalog No. AC384520010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (ex F)
Physical Form Crystalline Powder
Formula Weight 104.9
Quantity 1g
Color Red to brown
Packaging Plastic bottle
Identification Confirmed
Linear Formula TiF3
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula F3Ti
MDL Number MFCD00042544
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with acids liberates very toxic gas.
Chemical Name or Material Titanium(III) fluoride
Assay Percent Range 98%
CAS 13470-08-1
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Titanium(III) fluoride
Assay ≥97.5% (ex F)
Assay Percent Range 98%
Molecular Formula F3Ti
Linear Formula TiF3
Formula Weight 104.9
MDL Number MFCD00042544
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with acids liberates very toxic gas.

SDS & Safety Documents