missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(III) chloride, 20%w/v solution in 2N Hydrochloric acid,AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 429151000

Catalog No. AC429151000



Specifications

Specifications

Titanium(III) chloride
Cl3Ti
MFCD00011266
Titanous chloride
XJDNKRIXUMDJCW-UHFFFAOYSA-J
154.26
≈20% (TiCl3) (exact strength stated on certificate of analysis)
AcroSeal™ Glass Bottle
Purple
7705-07-9,7647-01-0,7732-18-5
TiCl3
15, 9637
Solubility in water: soluble. Other solubilities: soluble in alcohol,practically insoluble in ether
154.26
Liquid
1.2000g/mL
Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether
100mL

Chemical Identifiers

7705-07-9
154.26
XJDNKRIXUMDJCW-UHFFFAOYSA-J
Cl3Ti
MFCD00011266
Titanous chloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-728-9

SDS Documents