Learn More
Titanium(II) oxide pellets, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04294014
Description
Used for vacuum deposition. Also applications include sunscreens and UV-blocking in cosmetics, varnishes, wood preservatives, textile fibers, and packaging films. Also used as paint pigment, opacifying agent, welding rod fluxes, optical coatings, as a catalyst, ceramic finish coat, plastics, elastomers, coated fabrics, printing inks, roofing granules, glass, and in glazes. Also identified as Titanium monoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
>3000°C | |
1700°C | |
12137-20-1 | |
(metals basis) | |
MFCD00036279 | |
Insoluble in water. | |
O=[Ti] | |
63.87 | |
63.9 | |
Odorless |
Titanium (II) oxide pellets | |
8 to 9mm(0.32 to 0.36 in.) dia. x 6 to 7mm(0.24 to 0.28 in.) thick | |
25 g | |
OTi | |
titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis | |
OGIDPMRJRNCKJF-UHFFFAOYSA-N | |
oxotitanium | |
61685 | |
99.9% | |
4.95 g/mL |
Chemical Identifiers
12137-20-1 | |
63.87 | |
OGIDPMRJRNCKJF-UHFFFAOYSA-N | |
61685 | |
O=[Ti] |
OTi | |
MFCD00036279 | |
titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis | |
oxotitanium |
Safety and Handling
EINECSNumber : 235-236-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only