missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(II) Oxide, 99.5% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 7712636

 View more versions of this product

Catalog No. AA7712636



Specifications

Specifications

Titanium(II) Oxide
99.5%, metal basis
MFCD00036279
OGIDPMRJRNCKJF-UHFFFAOYSA-N
oxotitanium
61685
500g
12137-20-1
OTi
titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis
O=[Ti]
63.866
63.9

Chemical Identifiers

12137-20-1
63.866
OGIDPMRJRNCKJF-UHFFFAOYSA-N
61685
O=[Ti]
OTi
MFCD00036279
titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis
oxotitanium
SDS Documents