missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(II) hydride, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 12857D3

 View more versions of this product

Catalog No. AA12857D3



Specifications

Specifications

Titanium(II) hydride
99% (metals basis)
MFCD00151301
-325 Mesh Powder
2 x 1kg
7704-98-5
H2Ti
49.92
3.9g/mL

Chemical Identifiers

7704-98-5
H2Ti
MFCD00151301
Safety and Handling

Safety and Handling

EINECSNumber : 231-726-8

Documents