missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(II) Hydride, 99% (Metal Basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 8918330

 View more versions of this product

Catalog No. AA8918330



Specifications

Specifications

Titanium(II) Hydride
99%, metal basis
MFCD00151301
49.92
3.9
450°C (decomposition)
7704-98-5
H2Ti
Soluble in water.
1-3 Micron Powder
Air and moisture sensitive
250g

Chemical Identifiers

7704-98-5
H2Ti
MFCD00151301
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

P210-P240-P241-P280-P370+P378q

H228-H500

DOTInformation : DOT Class: 4.1; Packaging Group: II

EINECSNumber : 231-726-8

RTECSNumber : XR2130000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents