missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium ICP Standard, 1mL = 1mg Ti (1,000ppm Ti), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPTI1KW500

 View more versions of this product

Catalog No. PTI1KW-500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Titanium ICP Standard
98.0,0.4,0.01
Liquid
1,000ppm Ti
Colorless
16962-40-6,7664-39-3,7732-18-5
100.0,0.42,0.01
Poly natural
1 mL = 1 mg Ti
500mL

Chemical Identifiers

16962-40-6
Documents