Please login to your online account to display your discounted pricing

Titanium ICP Standard, 1mL = 10mg Ti (10,000ppm Ti), Ricca Chemical


Quantity 50mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIPTI10KW50

 View more versions of this product

Catalog No. PTI10KW-50

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 50mL
Equivalence 1 mL = 10 mg Ti
Chemical Name or Material Titanium ICP Standard
CAS 7732-18-5
CAS Min % 94.0
Concentration or Composition Notes 10,000ppm Ti
CAS Max % 98.0
CAS 7664-39-3
Packaging Poly natural
CAS Max % 4.05
CAS Min % 0.09
CAS Min % 3.89
CAS Max % 0.1
CAS 16962-40-6

Specifications

Chemical Name or Material Titanium ICP Standard
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 98.0
CAS 16962-40-6
CAS Min % 3.89
CAS Max % 4.05
CAS 7664-39-3
CAS Min % 0.09
CAS Max % 0.1
Physical Form Liquid