missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium Granules, -15 mesh, 99.9% (Metals basis excluding Na and K), Alfa Aesar™

Manufacturer:  Alfa Aesar 1399709

 View more versions of this product

Catalog No. AA1399709



Specifications

Specifications

Titanium
7440-32-6
99.9% (Metals basis excluding Na and K)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
Gray
10g
Dark Gray granules
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
Odorless
23963
Granular

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Documents