missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium gauze, 80 mesh woven from 0.13mm (0.005 in.) dia. wire

Manufacturer:  Alfa Aesar™ 46799GT

 View more versions of this product

Catalog No. AA46799GT



Specifications

Specifications

Titanium gauze
80 mesh woven from 0.13mm (0.005 in.) dia. wire
Ti
80 Mesh
RTAQQCXQSZGOHL-UHFFFAOYSA-N
[Ti]
47.867
CHEBI:33341
150 x 150mm
7440-32-6
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
Odorless
titanium
23963
Wire Cloth

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents