missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium Gauze, 18 mesh woven from 0.28mm (0.011 in.) dia. wire, , Alfa Aesar™

Manufacturer:  Alfa Aesar 43794GN

 View more versions of this product

Catalog No. AA43794GN



Specifications

Specifications

Titanium
7440-32-6
Ti
alloy
[Ti]
Odorless
23963
Gauze
125 x 125mm
Silver Gray guaze
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
Silver to Gray

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Documents