missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium foil, 99.5% (metals basis), Alfa Aesar™

Form: approx. 11.3g/100x100mm

Manufacturer:  Thermo Fisher Scientific 10385HY

 View more versions of this product

Catalog No. AA10385HY



Specifications

Specifications

Titanium
Annealed
Silver Gray foil
Ti
alloy, c.p., contimet 30, cp, ii hydride, jistp28, oremet, titan vt 1-1, titanium-125, vt1
[Ti]
Odorless
23963
Foil, 0.25mm (0.01 in.) thick
100 x 2000mm
7440-32-6
99.5% (Metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
Silver to Gray

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy, c.p., contimet 30, cp, ii hydride, jistp28, oremet, titan vt 1-1, titanium-125, vt1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

EINECSNumber : 231-142-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents