missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium foil, 99.5% (metals basis), Alfa Aesar™

Form: approx. 11.3g/100x100mm

Manufacturer:  Thermo Fisher Scientific 10385HP

 View more versions of this product

Catalog No. AA10385HP



Specifications

Specifications

Titanium
Annealed
Silver Gray foil
Ti
alloy
[Ti]
Odorless
23963
Foil, 0.25mm (0.01 in.) thick
100 x 500mm
7440-32-6
99.5% (Metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
Silver to Gray

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

EINECSNumber : 231-142-3

Documents