missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium foil, 2.0mm (0.08 in.) thick, annealed, 99.2% (metals basis)

Manufacturer:  Alfa Aesar™ 10398GH

 View more versions of this product

Catalog No. AA10398GH



Specifications

Specifications

Titanium foil
100 x 100mm
7440-32-6
Ti
alloy
[Ti]
Odorless
23963
Foil
≈90g/100x100mm
2.0mm (0.08 in.) thick, Annealed
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
99.2%

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents