missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium foil, 1.6mm (0.063 in.) thick, annealed, 99.2% (metals basis)

Manufacturer:  Alfa Aesar™ 44485GW

 View more versions of this product

Catalog No. AA44485GW



Specifications

Specifications

Titanium foil
200 x 200mm
7440-32-6
Ti
alloy
[Ti]
Odorless
23963
Foil
≈72g/100x100mm
1.6mm (0.063 in.) thick, Annealed
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
99.2%

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

EINECSNumber : 231-142-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents