missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium foil, 0.5mm (0.02 in.) thick, stress relieved, 99.99+% (metals basis)

Manufacturer:  Alfa Aesar™ 43676FI

 View more versions of this product

Catalog No. AA43676FI



Specifications

Specifications

Titanium foil
50 x 50mm
7440-32-6
Ti
alloy
[Ti]
Odorless
23963
Foil
≈1.4g/25x25mm
0.5mm (0.02 in.) thick, stress relieved
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
≥99.99%

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents