missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium foil, 0.127mm (0.005 in.) thick, annealed, 99.99+% (metals basis)

Manufacturer:  Alfa Aesar™ 45584GH

 View more versions of this product

Catalog No. AA45584GH



Specifications

Specifications

Titanium foil
0.127mm (0.005 in.) thick, Annealed
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
[Ti]
47.867
CHEBI:33341
≥99.99%
100 x 100mm
7440-32-6
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
Odorless
titanium
23963
Foil

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents