missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium foil, 0.05mm (0.002 in.) thick, annealed, Grade 1, 99.6% (metals basis)

Manufacturer:  Alfa Aesar™ 44202GT

 View more versions of this product

Catalog No. AA44202GT



Specifications

Specifications

Titanium foil, Grade 1
150 x 150mm
7440-32-6
Ti
alloy
[Ti]
Odorless
23963
Foil
Grade 1
≈0.14g/25x25mm
0.05mm (0.002 in.) thick, annealed, Temper: soft
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
99.6%

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

EINECSNumber : 231-142-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents