missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol

Manufacturer:  Alfa Aesar™ 8936036

 View more versions of this product

Catalog No. AA8936036



Specifications

Specifications

Titanium (diisopropoxide) bis (2,4-pentanedionate)
C16H28O6Ti
UN1219
364.3
75% in isopropanol
Alcohol-like
Moisture Sensitive
500g
17927-72-9
MFCD00000031
Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV)
Liquid
1.010g/mL
1.494
Brown
18°C (61°F)

Chemical Identifiers

17927-72-9
C16H28O6Ti
MFCD00000031
Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV)
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL

TSCA : TSCA

Documents