Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium cube, 6.4x6.4x6.4mm (0.25x0.25x0.25 in.), 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Titanium cube
CAS 7440-32-6
Name Note 6.4x6.4x6.4mm (0.25x0.25x0.25 in.)
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13998QJ

 
alfa aesar™
13998QJ
5 slug CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13998QI

 
alfa aesar™
13998QI
1 slug CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13998QK

 
alfa aesar™
13998QK
25 slug CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Cubes
Chemical Name or Material Titanium cube
TSCA TSCA
Name Note 6.4x6.4x6.4mm (0.25x0.25x0.25 in.)
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00011264
CAS 7440-32-6
Odor Odorless
Weight ≈1.2g/cube
Quantity 5 slug