missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium cube, 6.4x6.4x6.4mm (0.25x0.25x0.25 in.), 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 13998QK

 View more versions of this product

Catalog No. AA13998QK



Specifications

Specifications

Titanium cube
25 slug
7440-32-6
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
Odorless
23963
Cubes
≈1.2g/cube
6.4x6.4x6.4mm (0.25x0.25x0.25 in.)
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
99.99%

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents