Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium carbide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis)

Magnetron sputtering source

$587.64 - $587.64

Chemical Identifiers

Chemical Name or Material Titanium carbide sputtering target
CAS 12070-08-5
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 59.91
MDL Number MFCD00011268
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41106KS

  SDS
alfa aesar™
41106KS
1 each Each for $587.64
Description & Specifications

Specifications

Formula Weight 59.91
Assay Percent Notes (metals basis)
Quantity 1 each
Chemical Name or Material Titanium carbide sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.5%
MDL Number MFCD00011268
CAS 12070-08-5
RTECS Number XR1903500