Titanium carbide, Alfa Aesar™
Manufacturer:  Alfa Aesar 4492222
 View more versions of this product
Catalog No. AA4492222
Specifications
Specifications
Titanium carbide
CTi
59.91
100g
12070-08-5
MFCD00011268
30 To 50nm APS Powder, S.A. 35 To 45m2/g
Chemical Identifiers
12070-08-5
CTi
MFCD00011268
Safety and Handling
Safety and Handling
EINECSNumber : 235-120-4
SDS Documents