missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium carbide, 99.5% (metals basis), Alfa Aesar™

$86.46 - $699.14

Chemical Identifiers

CAS 12070-08-5
Molecular Formula CTi
MDL Number MFCD00011268
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4017818
View Documents
Thermo Fisher Scientific
4017818
50g Each for $86.46
 
AA4017830
View Documents
Thermo Fisher Scientific
4017830
250g Each for $210.54
 
AA40178A1
View Documents
Thermo Fisher Scientific
40178A1
1kg Each for $699.14
 
Specifications

Chemical Identifiers

12070-08-5
CTi
MFCD00011268

Specifications

12070-08-5
CTi
Insoluble in water.
Typically 2 Micron Powder
4820°C
50g
99.5% (metals basis)
MFCD00011268
59.91
4.93 g/mL
∽3140°C
Titanium carbide
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 235-120-4

RTECSNumber : XR1903500

TSCA : Yes

Recommended Storage : Ambient temperatures