Please login to your online account to display your discounted pricing

Titanium, -100 Mesh, 99.5%, Powder, ACROS Organics™


Quantity 500g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC421015000

 View more versions of this product

Catalog No. AC421015000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Titanium
Quantity 500g
Name Note -100 Mesh
CAS 7440-32-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Flammable solid.
Catches fire spontaneously if exposed to air.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
Wear face protection.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use sand for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Density 4.5100g/mL
Physical Form Fine Powder
Assay Percent Range 99.5%
Molecular Formula Ti
Linear Formula Ti
Formula Weight 47.88
MDL Number MFCD00011264
Fieser 07,368; 08,481; 09,466; 11,526; 15,315
Merck Index 15, 9626
Packaging Glass Bottle
Total Trace Metal Impurities 0.5% max.
Color Gray

Specifications

Chemical Name or Material Titanium
Name Note -100 Mesh
CAS 7440-32-6
Physical Form Fine Powder
Assay Percent Range 99.5%
Molecular Formula Ti
Linear Formula Ti
Formula Weight 47.88
MDL Number MFCD00011264
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
Wear face protection.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use sand for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Catches fire spontaneously if exposed to air.

SDS & Safety Documents