Please login to your online account to display your discounted pricing

Titanium, -100 Mesh, 99.5%, Powder, ACROS Organics™

$159.70 - $159.70

Chemical Identifiers

Chemical Name or Material Titanium
Name Note -100 Mesh
CAS 7440-32-6
Physical Form Fine Powder
Assay Percent Range 99.5%
Molecular Formula Ti
Linear Formula Ti
Formula Weight 47.88
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC421015000

  SDS
thermo fisher scientific
AC421015000
Glass Bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC421011000

  SDS
thermo fisher scientific
421011000
Glass Bottle 100g Each for $159.70

AC421010100

  SDS
thermo fisher scientific
AC421010100
Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Powder
Formula Weight 47.88
Quantity 500g
Color Gray
Packaging Glass Bottle
Density 4.5100g/mL
Fieser 07,368; 08,481; 09,466; 11,526; 15,315
Merck Index 15, 9626
Chemical Name or Material Titanium
Name Note -100 Mesh
Assay Percent Range 99.5%
Molecular Formula Ti
Linear Formula Ti
MDL Number MFCD00011264
CAS 7440-32-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Flammable solid.
Catches fire spontaneously if exposed to air.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
Wear face protection.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use sand for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Total Trace Metal Impurities 0.5% max.
Safety and Handling

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
Wear face protection.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use sand for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Catches fire spontaneously if exposed to air.