Please login to your online account to display your discounted pricing

Tiron, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC174145000

 View more versions of this product

Catalog No. AC174145000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 332.21
Quantity 500g
Color Off-white
Beilstein 11, 297
Merck Index 15, 9621
Packaging Glass bottle
Chemical Name or Material Tiron™
Molecular Formula C6H4Na2O8S2·H2O
MDL Number MFCD00149531
Synonym 4,5-Dihydroxy-1,3-benzenedisulfonic acid, disodium salt monohydrate
Linear Formula (HO)2C6H2(SO3Na)2·H2O
CAS 149-45-1
Infrared Spectrum Authentic
Loss on Drying 4% to 6% (typically)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tiron™
CAS 149-45-1
Molecular Formula C6H4Na2O8S2·H2O
Linear Formula (HO)2C6H2(SO3Na)2·H2O
Formula Weight 332.21
MDL Number MFCD00149531
Synonym 4,5-Dihydroxy-1,3-benzenedisulfonic acid, disodium salt monohydrate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents