Please login to your online account to display your discounted pricing

Thermo Scientific™ Automation Tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms with 384-Channel Disposable Tip Head


Increase pipetting performance with Thermo Scientific™ Tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.

Manufacturer: thermo scientific™  946262

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 02-681-063

  • / Pack of 3840 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Color Clear
Product Line Agilent Bravo
Volume (Metric) 10μL
For Use With (Equipment) Bravo and VPrep Automated Liquid Handling Platforms
Filtered No
Sterility Nonsterile
Packaging 384 Tips/Rack, 10 Racks/Case, 5 Packs/Case

Increase pipetting performance with Thermo Scientific™ Tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. The result is accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. These automation tips are validated on the actual workstations for which they are created, and are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.

  • Available in nonsterile and sterile
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)