Please login to your online account to display your discounted pricing

Tiotidine, Tocris


Potent, selective H2 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS082610

 View more versions of this product

Catalog No. 082610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 312.41
Chemical Name or Material Tiotidine
Molecular Formula C10H16N8S2
CAS 69014-14-8
Assay Percent Range >99%
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Tiotidine
Assay Percent Range >99%
Molecular Formula C10H16N8S2
Formula Weight 312.41
Accessories

Store at Room Temperature