Tingley™ Rubber Work High-top Galoshes
Won't crack or stiffen, even in cold weather
Manufacturer:  Tingley™ 1300XL
 View more versions of this product
Catalog No. 19-051-964
Description
Description
  • Made from an exclusive stretch-rubber compound
  • Protect against alcohol, and many organic acids, salts and alkalis
  • No hardware to tear out or conduct electricity
  • No lining for easy decontamination
  • Deep, slip-resistant treads
  • Black
Specifications
Specifications
Chemicals, acids, alcohols
Black
Rubber
Deep-cleated outsole
Button
Food processing, transportation, utilities, construction, general industry, municipalities, and agriculture
11 to 12.5,X-Large
High top galoshes