missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tingley™ Better Grade PVC Work Boots

Blend of high-quality PVC resins, low-temperature plasticizers, and nitrile rubber creates work boots that remain supple even in cold weather

$34.93 - $48.45

Specifications

Color Brown
For Use With (Application) Ideal for agriculture, industrial, and food processing applications.
Height (English) 15 in.
Height (Metric) 38cm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Material Size Price Quantity  
Catalog Number Mfr. No. Material Size Price Quantity  
19-022945 Tingley Rubber Corp
5114408
PVC; Plain toe 8 Pack for $34.93
19-022942 Tingley Rubber Corp
5114409
PVC; Plain toe 9 Pack for $34.93
19022939 Tingley Rubber Corp
51144
PVC; Plain toe 12 Pack for $36.02
19022938 Tingley Rubber Corp
005114411
PVC; Plain toe 11 Pack for $36.02
19022940 Tingley Rubber Corp
005114413
PVC; Plain toe 13 Pack for $36.02
19022937 Tingley Rubber Corp
005114410
PVC; Plain toe 10 Pack for $36.53
19022946 Tingley Rubber Co
005124410
PVC; Steel toe 10 Pack for $48.45
Specifications

Description

Withstand abrasion, and provide resistance to chemicals such as alcohols, organic and inorganic acids, salts, and alkalis. 100% waterproof. Ideal for agriculture, industrial, and food processing applications.

  • 15 in. high (38cm)
  • Brown upper with crepe sole
  • Cleated outsole spits out debris for excellent gripping action
  • Injection molded construction for 100% waterproof protection
  • Flexible upper material stays supple in cold temperatures to make walking easier
  • Outsole material is soft for excellent slip resistance and absorbs shock when walking
  • Removable polypropylene insole wicks perspiration to help keep feet dry and comfortable

Compliance

Steel-toe work boots meet ANSI Z41 PT99 M/I/75/C/75.

Description & Specifications

Specifications

Brown
Ideal for agriculture, industrial, and food processing applications.
15 in.
38cm