missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin wire, 99.998% (metals basis), Alfa Aesar™, Puratronic™

Form: approx. 5.72g/m

Manufacturer:  Alfa Aesar 11905BW

 View more versions of this product

Catalog No. AA11905BW



Specifications

Specifications

Tin
7440-31-5
7.31g/cm3
Sn
flake
[Sn]
Odorless
5352426
Wire, 1.0mm (0.04 in.) dia.
50cm
Silver Gray wire
99.998% (Metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
Silver to Gray

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

EINECSNumber : 23-141-8

Documents