missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin wire, 99.95% (metals basis), Alfa Aesar™

Form: approx. 58g/m

Manufacturer:  Alfa Aesar 10790G5

 View more versions of this product

Catalog No. AA10790G5



Specifications

Specifications

Tin
7440-31-5
99.95% (Metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
Silver to Gray
5m
Silver Gray wire
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
[Sn]
Odorless
5352426
Wire, 3.2mm (0.13 in.) dia.

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

EINECSNumber : 23-141-8

Documents