missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin Wire, 2.0mm (0.08 in.) dia., Puratronic™, 99.9985% (Metals basis), Alfa Aesar™

Form: Approx. 22.9g/m

Manufacturer:  Alfa Aesar 11904CC

 View more versions of this product

Catalog No. AA11904CC



Specifications

Specifications

Tin
7440-31-5
99.9985% (Metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
Silver to Gray
500cm
Silver Gray wire
Sn
flake
[Sn]
Odorless
5352426
Wire, 2mm (0.08 in.) dia.

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake
CHEBI:32990
[Sn]
Documents