Please login to your online account to display your discounted pricing

Tin Standard 1000mg/L in 20% HCL (Assurance Grade), SPEX CertiPrep

$83.09 - $237.91

Chemical Identifiers

Chemical Name or Material Tin Standard
CAS 7440-31-5
CAS Min % 1.0
CAS Max % 1.0
CAS 7647-01-0
CAS Min % 20.0
CAS Max % 20.0
CAS 7732-18-5
CAS Min % 79.0
CAS Max % 79.0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PLSN52T

 
spex certiprep llc
PLSN52T
PTFE 250mL Each for $237.91

PLSN52M

 
spex certiprep llc
SPXPLSN52M
Triple-leached LDPE bottle 30mL Each for $83.09
Description & Specifications

Specifications

Quantity 250mL
Recommended Storage Ambient
DOT Information Nonhazardous
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Chemical Name or Material Tin Standard
CAS Max % 79.0
CAS 7440-31-5
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
1% HNO3/1% HF    1000μg/mL    PLSN2-2X
2% HNO3/2% HF    10,000    PLSN2-3X
20% HCl    1000μg/mL    PLSN5-2Y, -2T, -2X    10,000    PLSN5-3Y, -3X
CAS Min % 1.0
CAS Max % 1.0
CAS 7647-01-0
Packaging PTFE
CAS Min % 20.0
CAS Max % 20.0
CAS 7732-18-5
CAS Min % 79.0

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Ambient