Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.995% (metals basis)

$574.08 - $574.08

Chemical Identifiers

Chemical Name or Material Tin sputtering target
CAS 7440-31-5
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00133862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40900KS

  SDS
alfa aesar™
40900KS
1 each Each for $574.08
Description & Specifications

Specifications

Assay 99.995%
Odor Odorless
MDL Number MFCD00133862
Quantity 1 each
Assay Percent Notes (metals basis)
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Chemical Name or Material Tin sputtering target
TSCA TSCA
CAS 7440-31-5